CAS RN 16924-03-1
IUPAC Name:
niobium(5+) dipotassium heptafluoride
Synonyms:
niobium(5+) dipotassium heptafluoride
Molecular Weight (g/mol):
304.09
Molecular Formula:
F7K2Nb
InChi Key:
FTPUNAWAGWERLA-UHFFFAOYSA-G
SMILES:
[F-].[F-].[F-].[F-].[F-].[F-].[F-].[K+].[K+].[Nb+5]
1
–
1
of
1
results
1
Potassium heptafluoroniobate(V), 99.5%
CAS: 16924-03-1 Molecular Formula: K2NbF7 MDL Number: MFCD00042368