accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16925-25-0

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 258.74
Molecular Formula: F6NaSb
InChi Key: ZJLVIRKMUXZEDU-UHFFFAOYSA-N
SMILES: [F].[F].[F].[F].[F].[F].[Na+].[Sb]
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (3)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (2)
  • (6)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)