accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C(S)NHO(S)HOOHNO(S)OHONHH3COOHN(S)OHOO

CAS RN 169332-60-9

IUPAC Name: (4S)-4-[(2S)-3-carboxy-2-acetamidopropanamido]-4-{[(1S)-1-{[(2S)-1-carboxy-3-oxopropan-2-yl]carbamoyl}-2-methylpropyl]carbamoyl}butanoic acid
Synonyms: ac-asp-glu-val-asp-H
Molecular Weight (g/mol): 502.48
Molecular Formula: C20H30N4O11
InChi Key: UMBVAPCONCILTL-MRHIQRDNSA-N
SMILES: CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C=O
Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results