CAS RN 16937-92-1
CAS RN 16937-92-1
IUPAC Name:
5-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms:
5-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol):
366.41
Molecular Formula:
C18H26N2O6
InChi Key:
QYYCZJUFHDLLOJ-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
Ndelta-Benzyloxycarbonyl-Nalpha-Boc-D-ornithine, 98%, Thermo Scientific™
CAS: 16937-92-1 Molecular Formula: C18H26N2O6 Molecular Weight (g/mol): 366.41 MDL Number: MFCD00069970 InChI Key: QYYCZJUFHDLLOJ-UHFFFAOYNA-N Synonym: boc-d-orn z-oh,nalpha-boc-ndelta-cbz-r-ornithine,boc-d-orn z-oh tlc,2r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid,r-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid,n-boc-n'-cbz-d-ornithine,a-boc-n,ksc918o2r,a-z-n,a-boc-n,a-z-d-ornithine,a-z-n,a-boc-d-ornithine PubChem CID: 7272030 SMILES: CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(O)=O