accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHONHOOCH3CH3H3CH2O

CAS RN 16937-99-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid hydrate
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-methylpentanoic acid hydrate
Molecular Weight (g/mol): 249.31
Molecular Formula: C11H23NO5
InChi Key: URQQEIOTRWJXBA-UHFFFAOYNA-N
SMILES: O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)