accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16940-97-9

IUPAC Name: dipotassium hexachlororheniumdiuide
Synonyms: dipotassium hexachlororheniumdiuide
Molecular Weight (g/mol): 477.1
Molecular Formula: Cl6K2Re
InChi Key: HGCRXIXEXSEJGV-UHFFFAOYSA-H
SMILES: [K+].[K+].Cl[Re--](Cl)(Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)