accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16941-11-0

IUPAC Name: ammonium hexafluoro-λ⁵-phosphanuide
Synonyms: ammonium hexafluorophosphate
Molecular Weight (g/mol): 163
Molecular Formula: F6H4NP
InChi Key: NIZXKAYXSNUDOU-UHFFFAOYSA-O
SMILES: [NH4+].F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol) 
  • (8)
  • (3)
  • (2)
Quantity 
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)
Percent Purity 
  • (4)
  • (4)
Physical Form 
  • (4)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (6)
  • (8)
  • (3)
  • (2)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (4)