accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNOOH3CCH3H3C

CAS RN 169447-70-5

IUPAC Name: tert-butyl 2-methylpiperazine-1-carboxylate
Synonyms: tert-butyl 2-methylpiperazine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: DATRVIMZZZVHMP-UHFFFAOYNA-N
SMILES: CC1CNCCN1C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results