accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169530-97-6

IUPAC Name: 2-amino-3-(3-nitrophenyl)propanoic acid
Synonyms: 2-amino-3-(3-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: YTHDRUZHNYKZGF-UHFFFAOYNA-N
SMILES: NC(CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)