CAS RN 169565-17-7
CAS RN 169565-17-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ (±)-Amphetamine-d11 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Suitable for quantitation of amphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or sports testing.