accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSiFFFFFHH

CAS RN 16961-83-4

IUPAC Name: dihydrogen hexafluorosilanediuide
Synonyms: dihydrogen hexafluorosilanediuide
Molecular Weight (g/mol): 144.09
Molecular Formula: F6H2Si
InChi Key: OHORFAFFMDIQRR-UHFFFAOYSA-P
SMILES: [H+].[H+].F[Si--](F)(F)(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)