accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4FHGeH4FHFHHFFF

CAS RN 16962-47-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 230.76
Molecular Formula: F6GeH16N2
InChi Key: LHKQHMLGVWLMTF-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].F.F.F.F.[F-].[F-].[GeH4]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results