accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169685-34-1

IUPAC Name: N4,N4'-bis(4-methoxy-2-methylphenyl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4'-bis(4-methoxy-2-methylphenyl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 576.74
Molecular Formula: C40H36N2O2
InChi Key: HACXFZALVVMGPO-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=C(C)C=C(OC)C=C1
Percent Purity 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results