accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNBr

CAS RN 1698-16-4

IUPAC Name: 10-bromo-7H-benzo[c]carbazole
Synonyms: 10-bromo-7H-benzo[c]carbazole
Molecular Weight (g/mol): 296.17
Molecular Formula: C16H10BrN
InChi Key: YHAHNQXQOZYZLP-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC3=C2C2=CC=CC=C2C=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results