accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16982-21-1

IUPAC Name: ethyl carbamothioylformate
Synonyms: ethyl carbamothioylformate
Molecular Weight (g/mol): 133.17
Molecular Formula: C4H7NO2S
InChi Key: YMBMCMOZIGSBOA-UHFFFAOYSA-N
SMILES: CCOC(=O)C(N)=S
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)