accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNClNN

CAS RN 1700-02-3

IUPAC Name: 2,4-dichloro-6-phenyl-1,3,5-triazine
Synonyms: 2,4-dichloro-6-phenyl-1,3,5-triazine
Molecular Weight (g/mol): 226.06
Molecular Formula: C9H5Cl2N3
InChi Key: AMEVJOWOWQPPJQ-UHFFFAOYSA-N
SMILES: ClC1=NC(=NC(Cl)=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)