accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNH2N

CAS RN 170017-74-0

IUPAC Name: tert-butyl 4-(2-aminophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(2-aminophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 277.37
Molecular Formula: C15H23N3O2
InChi Key: LNDQGWAWYKFYAO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results