accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1701-93-5

IUPAC Name: silver(1+) cyanosulfanide
Synonyms: silver thiocyanate
Molecular Weight (g/mol): 165.95
Molecular Formula: CAgNS
InChi Key: RHUVFRWZKMEWNS-UHFFFAOYSA-M
SMILES: [Ag+].[S-]C#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results