accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH2COCH2

CAS RN 17018-07-4

IUPAC Name: 1,3-bis(prop-2-en-1-yloxy)propan-2-ol
Synonyms: 1,3-bis(prop-2-en-1-yloxy)propan-2-ol
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: OFLGYLDXUIXHQY-UHFFFAOYSA-N
SMILES: OC(COCC=C)COCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results