accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2

CAS RN 17027-51-9

IUPAC Name: 2-{[1,1'-biphenyl]-4-yl}ethan-1-amine
Synonyms: 2-{[1,1'-biphenyl]-4-yl}ethanamine
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: WHPLBPSDTIZFSX-UHFFFAOYSA-N
SMILES: NCCC1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results