accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNOH

CAS RN 170299-61-3

IUPAC Name: tert-butyl N-[2-(hydroxymethyl)cyclopropyl]carbamate
Synonyms: tert-butyl N-[2-(hydroxymethyl)cyclopropyl]carbamate
Molecular Weight (g/mol): 187.24
Molecular Formula: C9H17NO3
InChi Key: OCKKMJSVLVALMF-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CC1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)