accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1703-46-4

IUPAC Name: [4-(dimethylamino)phenyl]methanol
Synonyms: [4-(dimethylamino)phenyl]methanol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: WQBCAASPALGAKX-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(CO)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results