accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OOH

CAS RN 1704-62-7

IUPAC Name: 2-[2-(dimethylamino)ethoxy]ethan-1-ol
Synonyms: 2-[2-(dimethylamino)ethoxy]ethanol
Molecular Weight (g/mol): 133.19
Molecular Formula: C6H15NO2
InChi Key: YSAANLSYLSUVHB-UHFFFAOYSA-N
SMILES: CN(C)CCOCCO
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)