accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOCH3OOCH3OOCH3OOH3C

CAS RN 17042-40-9

IUPAC Name: [3,4,5-tris(acetyloxy)-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 454.43
Molecular Formula: C21H26O11
InChi Key: RPHXBVOPPUTUES-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C2OC(C)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results