accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3MgBr

CAS RN 17049-49-9

IUPAC Name: bromo(octyl)magnesium
Synonyms: bromo(octyl)magnesium
Molecular Weight (g/mol): 217.43
Molecular Formula: C8H17BrMg
InChi Key: ZDUZWGUHIRQPGP-UHFFFAOYSA-M
SMILES: CCCCCCCC[Mg]Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results