accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3H3C

CAS RN 1706-11-2

IUPAC Name: 2-methoxy-1,4-dimethylbenzene
Synonyms: 1,4-dimethyl-2-methyoxybenzene
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: SJZAUIVYZWPNAS-UHFFFAOYSA-N
SMILES: COC1=CC(C)=CC=C1C
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results