accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOOHHOOH3CH3CO

CAS RN 1707-77-3

IUPAC Name: 1,2-bis(2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Synonyms: 1,2-bis(2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Molecular Weight (g/mol): 262.3
Molecular Formula: C12H22O6
InChi Key: ODYBCPSCYHAGHA-UHFFFAOYNA-N
SMILES: CC1(C)OCC(O1)C(O)C(O)C1COC(C)(C)O1
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)