accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOCl

CAS RN 17070-58-5

IUPAC Name: 1-benzofuran-2-sulfonyl chloride
Synonyms: 1-benzofuran-2-sulfonyl chloride
Molecular Weight (g/mol): 216.64
Molecular Formula: C8H5ClO3S
InChi Key: RWCKBBSBRTUUHR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)