accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 170737-46-9

IUPAC Name: 6-bromonaphthalene-2-carbaldehyde
Synonyms: 6-bromonaphthalene-2-carbaldehyde
Molecular Weight (g/mol): 235.08
Molecular Formula: C11H7BrO
InChi Key: DLLDUYJRQNTEOR-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=C(C=O)C=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results