accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17078-27-2

IUPAC Name: bis[4-(dimethylamino)phenyl]ethane-1,2-dione
Synonyms: bis[4-(dimethylamino)phenyl]ethane-1,2-dione
Molecular Weight (g/mol): 296.37
Molecular Formula: C18H20N2O2
InChi Key: AVFUVYIDYFXFSX-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)