accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 170787-99-2

IUPAC Name: sodium 2-(4-{[(3,5-dimethylphenyl)carbamoyl]methyl}phenoxy)-2-methylpropanoate
Synonyms: sodium 2-(4-{[(3,5-dimethylphenyl)carbamoyl]methyl}phenoxy)-2-methylpropanoate
Molecular Weight (g/mol): 363.39
Molecular Formula: C20H22NNaO4
InChi Key: SWDPIHPGORBMFR-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC(NC(=O)CC2=CC=C(OC(C)(C)C([O-])=O)C=C2)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results