accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1708-40-3

OHOO

CAS RN 1708-40-3

IUPAC Name: 2-phenyl-1,3-dioxan-5-ol
Synonyms: 2-phenyl-1,3-dioxan-5-ol
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: BWKDAAFSXYPQOS-UHFFFAOYSA-N
SMILES: OC1COC(OC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)