accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFFH2N

CAS RN 170804-18-9

IUPAC Name: ethyl 3-amino-4,4,4-trifluorobutanoate
Synonyms: ethyl 3-amino-4,4,4-trifluorobutanoate
Molecular Weight (g/mol): 185.15
Molecular Formula: C6H10F3NO2
InChi Key: KGUOZZWGWBBRRF-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)