accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3COSiOSiCH3CH3OSiH3CCH3OSiCH3H3C

CAS RN 17082-47-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 324.7
Molecular Formula: C8H24O4Si5
InChi Key: UOUILILVWRHZSH-UHFFFAOYSA-N
SMILES: C[Si](C)O[Si](O[Si](C)C)(O[Si](C)C)O[Si](C)C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)