accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCH3NH3CH3CH3C

CAS RN 17083-85-1

IUPAC Name: tetraethylazanium boron
Synonyms: tetraethylammonium boron
Molecular Weight (g/mol): 141.06
Molecular Formula: C8H20BN
InChi Key: NQZKZGHOYUYCHU-UHFFFAOYSA-N
SMILES: [B].CC[N+](CC)(CC)CC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results