accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3(E)(E)

CAS RN 17088-28-7

IUPAC Name: ethyl (2E)-3-{4-[(1E)-3-ethoxy-3-oxoprop-1-en-1-yl]phenyl}prop-2-enoate
Synonyms: ethyl (2E)-3-{4-[(1E)-3-ethoxy-3-oxoprop-1-en-1-yl]phenyl}prop-2-enoate
Molecular Weight (g/mol): 274.32
Molecular Formula: C16H18O4
InChi Key: QYGWZBFQWUBYAT-WGDLNXRISA-N
SMILES: CCOC(=O)\C=C\C1=CC=C(\C=C\C(=O)OCC)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results