accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOFFF

CAS RN 170886-13-2

IUPAC Name: 2-(trifluoromethyl)-1,4-dihydropyridin-4-one
Synonyms: 2-(trifluoromethyl)-1H-pyridin-4-one
Molecular Weight (g/mol): 163.1
Molecular Formula: C6H4F3NO
InChi Key: LKHCWMQMRXAPHX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)C=CN1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)