accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1709-59-7

IUPAC Name: 4-amino-N,N-dimethylbenzene-1-sulfonamide
Synonyms: N1-dimethylsulfanilamide
Molecular Weight (g/mol): 200.26
Molecular Formula: C8H12N2O2S
InChi Key: BABGMPQXLCJMSK-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results