CAS RN 170930-46-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Arp2/3 Complex Inhibitor I, Inactive Control, CK-689
The Arp2/3 Complex Inhibitor I, Inactive Control, CK-689, also referenced under CAS 170930-46-8, controls the biological activity of Arp2/3. This small molecule/inhibitor is primarily used for Cell Structure applications.