accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCH3FHOOH

CAS RN 170981-26-7

IUPAC Name: (2-fluoro-4-methylphenyl)boronic acid
Synonyms: 2-fluoro-4-methylphenylboronic acid
Molecular Weight (g/mol): 153.95
Molecular Formula: C7H8BFO2
InChi Key: LIXXGOMAGHXIMP-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(C=C1)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results