accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOH3CFFHOOH

CAS RN 170981-41-6

IUPAC Name: (2,3-difluoro-4-methoxyphenyl)boronic acid
Synonyms: 2,3-difluoro-4-methoxyphenylboronic acid
Molecular Weight (g/mol): 187.94
Molecular Formula: C7H7BF2O3
InChi Key: HIWYFMBSFSTYGU-UHFFFAOYSA-N
SMILES: COC1=C(F)C(F)=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results