accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH(E)(E)

CAS RN 17102-64-6

IUPAC Name: (2E,4E)-hexa-2,4-dien-1-ol
Synonyms: 2,4-hexadienol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: MEIRRNXMZYDVDW-MQQKCMAXSA-N
SMILES: C\C=C\C=C\CO
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)