accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1711-10-0

IUPAC Name: 3-iodobenzoyl chloride
Synonyms: 3-iodobenzoyl chloride
Molecular Weight (g/mol): 266.46
Molecular Formula: C7H4ClIO
InChi Key: DTELTOREECFDBC-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(I)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results