accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17114-97-5

IUPAC Name: methyl 2-amino-3-(tert-butoxy)propanoate hydrochloride
Synonyms: methyl 2-amino-3-(tert-butoxy)propanoate hydrochloride
Molecular Weight (g/mol): 211.69
Molecular Formula: C8H18ClNO3
InChi Key: PCIABNBULSRKSU-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)COC(C)(C)C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)