accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2(S)OHCH3CH2

CAS RN 17123-63-6

IUPAC Name: (3S)-2,5-dimethylhexa-1,5-dien-3-ol
Synonyms: (3S)-2,5-dimethylhexa-1,5-dien-3-ol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: QQMFDURNWCUKNS-QMMMGPOBSA-N
SMILES: CC(=C)C[C@H](O)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)