CAS RN 171235-71-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ TAPI-1
TAPI-1, CAS 171235-71-5, is a structural analog of TAPI-0 with similar in vitro efficacy for the inhibition of MMPs and TACE. Blocks the shedding of several cell surface proteins.