accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COBOH3CH3CNOO

CAS RN 171364-83-3

IUPAC Name: 4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 249.07
Molecular Formula: C12H16BNO4
InChi Key: LUWACRUAJXZANC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results