accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 171364-86-6

IUPAC Name: 2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(1-benzothiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 260.16
Molecular Formula: C14H17BO2S
InChi Key: LTSGSDOTQABJMA-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results