accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17138-28-2

IUPAC Name: ethyl 2-(4-hydroxyphenyl)acetate
Synonyms: ethyl 2-(4-hydroxyphenyl)acetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: HYUPPKVFCGIMDB-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(O)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)