accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 1714-29-0

IUPAC Name: 1-bromopyrene
Synonyms: 1-bromopyrene
Molecular Weight (g/mol): 281.15
Molecular Formula: C16H9Br
InChi Key: HYGLETVERPVXOS-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)